Learn More
Ace Glass, Inc. Adapter, SS, 1.5in beaded pipe to 1.5in female NPT
SDP
Catalog No. 50976627
Encompass
304 stainless steel adapter for matching 1.5 glass beaded process pipe to 1.5 female NPT fittings, gauges, and flanges.
Catalog No. 50-976-627 Supplier Ace Glass, Inc. Supplier No. 887124
304 stainless steel adapter for matching 1.5 glass beaded process pipe to 1.5 female NPT fittings, gauges, and flanges.