Learn More
Ace Glass, Inc. Adapter, SS, 1in beaded pipe to 1in male NPT
SDP
Catalog No. 50979947
Encompass
304 Stainless Steel adapter connects 1 beaded pipe to 1 male NPT fittings, gauges, and flanges.
Catalog No. 50-979-947 Supplier Ace Glass, Inc. Supplier No. 887006
304 Stainless Steel adapter connects 1 beaded pipe to 1 male NPT fittings, gauges, and flanges.