Learn More
Ace Glass, Inc. Adapter, SS, 1in beaded pipe to 1in female NPT
SDP
Catalog No. 50976626
Encompass
304 stainless steel adapter for matching 1 glass beaded process pipe to 1 female NPT fittings, gauges, and flanges.
Catalog No. 50-976-626 Supplier Ace Glass, Inc. Supplier No. 887122
304 stainless steel adapter for matching 1 glass beaded process pipe to 1 female NPT fittings, gauges, and flanges.