Learn More
Ace Glass, Inc. Adapter, SS, 2in beaded pipe to 2in female NPT
SDP
Catalog No. 50976628
Encompass
304 stainless steel adapter for matching 2 glass beaded process pipe to 2 female NPT fittings, gauges, and flanges.
Catalog No. 50-976-628 Supplier Ace Glass, Inc. Supplier No. 887128
304 stainless steel adapter for matching 2 glass beaded process pipe to 2 female NPT fittings, gauges, and flanges.