Learn More
Ace Glass, Inc. Adapter, SS, 2in beaded pipe to 2in male NPT
SDP
Catalog No. 50976624
Encompass
304 Stainless Steel adapter connects 2 beaded pipe to 2 male NPT fittings, gauges, and flanges.
Catalog No. 50-976-624 Supplier Ace Glass, Inc. Supplier No. 887010
304 Stainless Steel adapter connects 2 beaded pipe to 2 male NPT fittings, gauges, and flanges.