Learn More
Ace Glass, Inc. Adapter, SS, 3in beaded pipe to 3in male NPT
SDP
Supplier:  Ace Glass, Inc. 887012
Encompass
304 Stainless Steel adapter connects 3 beaded pipe to 2 male NPT fittings, gauges, and flanges.
Catalog No. 50-976-625