Learn More
Ace Glass, Inc. Adapter,stainless,nw25-3/4in elbow hose barb
SDP
Catalog No. 5000854770
Encompass
304 Stainless Steel NW flange adapters for use with circulators. Connections either terminate in M16x1, M24x1.5, or M30x1.5 male. There is no need for thread sealent or teflon tape. Connections only need to be hand tightened then given a 1/4 turn with a wrench. Compatabile with all circulator companies connections having matching female connectors.
Catalog No. 50-008-54770 Supplier Ace Glass, Inc. Supplier No. 1218852
304 Stainless Steel NW flange adapters for use with circulators. Connections either terminate in M16x1, M24x1.5, or M30x1.5 male. There is no need for thread sealent or teflon tape. Connections only need to be hand tightened then given a 1/4 turn with a wrench. Compatabile with all circulator companies connections having matching female connectors.