Learn More
Chemglass Life Sciences PTFE Standard Taper Adapter, 24/40 Outer, Center Neck, 1/2" Male NPT
SDP
Catalog No. 501531284
Encompass_Preferred
PTFE Standard Taper Adapter, 24/40 Outer, Center Neck, 1/2" Male NPT. For center and vertical side necks of the CG-1945-T-50 7-Neck PTFE Reaction Vessel lid. Image shown is fully assembled Reaction Vessel lid.
Catalog No. 50-153-1284 Supplier Chemglass Life Sciences Supplier No. CG1945T33
PTFE Standard Taper Adapter, 24/40 Outer, Center Neck, 1/2" Male NPT. For center and vertical side necks of the CG-1945-T-50 7-Neck PTFE Reaction Vessel lid. Image shown is fully assembled Reaction Vessel lid.