Learn More
Chemglass Life Sciences Adapter, Syringe Addition, 14/20 Joint Size, 72mm Height
SDP
Catalog No. CG103914
Encompass_Preferred
Adapter, Syringe Addition, 14/20 Joint Size, 72mm Height. Adapter is inserted between the flask and condenser in a distillation set up and is used for additions to reactions with a syringe pump without having to bend the needle or cannula. Supplied complete with one each CG-3022-05 rubber septa.
Catalog No. CG103914 Supplier Chemglass Life Sciences Supplier No. CG103914
Adapter, Syringe Addition, 14/20 Joint Size, 72mm Height. Adapter is inserted between the flask and condenser in a distillation set up and is used for additions to reactions with a syringe pump without having to bend the needle or cannula. Supplied complete with one each CG-3022-05 rubber septa.