Learn More
Chemglass Life Sciences Adapter, Syringe Needle, 24/40 Inner Joint, 80mm Height
SDP
Catalog No. CG103601
Encompass_Preferred
Adapter, Syringe Needle, 24/40 Inner Joint, 80mm Height. Inner standard taper joint with the opposite end tooled to fit a CG-3022-05 or CG-3022-22 rubber septa. Supplied complete with one each rubber septa.
Catalog No. CG103601 Supplier Chemglass Life Sciences Supplier No. CG103601
Adapter, Syringe Needle, 24/40 Inner Joint, 80mm Height. Inner standard taper joint with the opposite end tooled to fit a CG-3022-05 or CG-3022-22 rubber septa. Supplied complete with one each rubber septa.