Learn More
Chemglass Life Sciences Adapter, Thermometer, Inlet, 19/22 Inner Joint, 55mm Height
SDP
Catalog No. CG103819
Encompass_Preferred
Adapter, Thermometer, Inlet, 19/22 Inner Joint, 55mm Height. Adapter has a tubulation at the top for use with a CG-1040 rubber fitting. Fitting permits insertion of plain stem thermometers or bleed tubes. Supplied complete with one CG-1040 rubber fitting.
Catalog No. CG103819 Supplier Chemglass Life Sciences Supplier No. CG103819
Adapter, Thermometer, Inlet, 19/22 Inner Joint, 55mm Height. Adapter has a tubulation at the top for use with a CG-1040 rubber fitting. Fitting permits insertion of plain stem thermometers or bleed tubes. Supplied complete with one CG-1040 rubber fitting.