Learn More
Chemglass Life Sciences Adapter, PTFE, Thermometer, Inlet, 10/18 Inner Joint, Use with Joint Sizes 10/18-10/30
SDP
Catalog No. CG104804
Encompass_Preferred
Adapter, PTFE, Thermometer, Inlet, 10/18 Inner Joint, Use with Joint Sizes 10/18-10/30. Made entirely of PTFE, adapter has an inner joint of the listed size. The compression nut allows for vertical adjustment of thermometers or bleed tubes having an O.D. between 6 and 7mm. Supplied complete with one #108 Viton o-ring.
Catalog No. CG104804 Supplier Chemglass Life Sciences Supplier No. CG104804
Adapter, PTFE, Thermometer, Inlet, 10/18 Inner Joint, Use with Joint Sizes 10/18-10/30. Made entirely of PTFE, adapter has an inner joint of the listed size. The compression nut allows for vertical adjustment of thermometers or bleed tubes having an O.D. between 6 and 7mm. Supplied complete with one #108 Viton o-ring.