Learn More
Chemglass Life Sciences Adapter, PTFE, Thermometer, Inlet, 14/20 Inner Joint, Use with Joint Sizes 14/20-14/35
SDP
Catalog No. CG104805
Encompass_Preferred
Adapter, PTFE, Thermometer, Inlet, 14/20 Inner Joint, Use with Joint Sizes 14/20-14/35. Made entirely of PTFE, adapter has an inner joint of the listed size. The compression nut allows for vertical adjustment of thermometers or bleed tubes having an O.D. between 6 and 7mm. Supplied complete with one #108 Viton o-ring.
Catalog No. CG104805 Supplier Chemglass Life Sciences Supplier No. CG104805
Adapter, PTFE, Thermometer, Inlet, 14/20 Inner Joint, Use with Joint Sizes 14/20-14/35. Made entirely of PTFE, adapter has an inner joint of the listed size. The compression nut allows for vertical adjustment of thermometers or bleed tubes having an O.D. between 6 and 7mm. Supplied complete with one #108 Viton o-ring.