Learn More
Chemglass Life Sciences 14/20 Joint Size, 1/4" Thermometer Size, #108 Upper O-Ring, #013 Lower O-Ring
SDP
Catalog No. 501530991
Encompass_Preferred
14/20 Joint Size, 1/4" Thermometer Size, #108 Upper O-Ring, #013 Lower O-Ring. PTFE adapter for use with 1/4 thermocouples. Adapter has an outer Perfluoro o-ring to form a grease-free, airtight seal between the adapter and the listed size of outer joint. The compression nut works in conjunction with the upper Perfluoro o-ring to provide a airtight seal between the adapter and thermocouple. Adapter design permits use with CG-145 Keck clips.
Catalog No. 50-153-0991 Supplier Chemglass Life Sciences Supplier No. CG1048P03
14/20 Joint Size, 1/4" Thermometer Size, #108 Upper O-Ring, #013 Lower O-Ring. PTFE adapter for use with 1/4 thermocouples. Adapter has an outer Perfluoro o-ring to form a grease-free, airtight seal between the adapter and the listed size of outer joint. The compression nut works in conjunction with the upper Perfluoro o-ring to provide a airtight seal between the adapter and thermocouple. Adapter design permits use with CG-145 Keck clips.