Learn More
Ace Glass, Inc. Thermocouple adapter, 14/20, use with 1/8in diameter probes, complete with PTFE bushing and PTFE ferrule
SDP
Catalog No. 50971680
Encompass_Preferred
Ace-Thred to 14/20 standard taper Thermocouple adapter with a PTFE bushing and PTFE ferrule. Thermocouple adapter is designed specifically to hold a 1/8 diameter temperature probe, thermometer, or tubing.
Catalog No. 50-971-680 Supplier Ace Glass, Inc. Supplier No. 504102
Ace-Thred to 14/20 standard taper Thermocouple adapter with a PTFE bushing and PTFE ferrule. Thermocouple adapter is designed specifically to hold a 1/8 diameter temperature probe, thermometer, or tubing.