Learn More
Ace Glass, Inc. Thermocouple adapter, 14/23, use with 3.175mm probes diameter probes, complete with PTFE bushing and PTFE ferrule
SDP
Catalog No. 50978300
Encompass_Preferred
Ace-Thred to 14/23 standard taper Thermocouple adapter with a PTFE bushing and PTFE ferrule. Thermocouple adapter is designed specifically to hold a 3.175mm diameter temperature probe, thermometer, or tubing.
Catalog No. 50-978-300 Supplier Ace Glass, Inc. Supplier No. 5041114
Ace-Thred to 14/23 standard taper Thermocouple adapter with a PTFE bushing and PTFE ferrule. Thermocouple adapter is designed specifically to hold a 3.175mm diameter temperature probe, thermometer, or tubing.