Learn More
Ace Glass, Inc. Thermocouple adapter, 24/40, use with 1/4in diameter probes, complete with PTFE bushing and FETFE; O-rings
SDP
Catalog No. 50971681
Encompass_Preferred
Ace-Thred to 24/40 standard taper Thermocouple adapter with a PTFE bushing and FETFE o-rings. Thermocouple adapter is designed specifically to hold a 1/4 diameter temperature probe, thermometer, or tubing.
Catalog No. 50-971-681 Supplier Ace Glass, Inc. Supplier No. 504110
Ace-Thred to 24/40 standard taper Thermocouple adapter with a PTFE bushing and FETFE o-rings. Thermocouple adapter is designed specifically to hold a 1/4 diameter temperature probe, thermometer, or tubing.