Learn More
Chemglass Life Sciences 29/42 Joint Size, 1/8" Thermometer Size, #006 Upper O-Ring, #022 Lower O-Ring
SDP
Catalog No. 50872872
Encompass_Preferred
29/42 Joint Size, 1/8" Thermometer Size, #006 Upper O-Ring, #022 Lower O-Ring. PTFE adapter for use with either 1/8¿ or 1/4¿ thermocouples. Adapter has an outer Perfluoro o-ring to form a grease-free, airtight seal between the adapter and the listed size of outer joint. The compression nut works in conjunction with the upper Perfluoro o-ring to provide a airtight seal between the adapter and thermocouple. Adapter design permits use with CG-145 Keck clips.
Catalog No. 50-872-872 Supplier Chemglass Life Sciences Supplier No. CG1048P06
29/42 Joint Size, 1/8" Thermometer Size, #006 Upper O-Ring, #022 Lower O-Ring. PTFE adapter for use with either 1/8¿ or 1/4¿ thermocouples. Adapter has an outer Perfluoro o-ring to form a grease-free, airtight seal between the adapter and the listed size of outer joint. The compression nut works in conjunction with the upper Perfluoro o-ring to provide a airtight seal between the adapter and thermocouple. Adapter design permits use with CG-145 Keck clips.