Learn More
Chemglass Life Sciences Adapter, Thermometer, Inlet, 10° Offset, 29/26 Inner Joint
SDP
Catalog No. 501530990
Encompass_Preferred
Adapter, Thermometer, Inlet, 10¿ Offset, 29/26 Inner Joint. Adapter has a #7 Chem-Thread at the top for vacuum tight seal of plain stem thermometers or any other tube having an O.D. between 4 and 7mm. When used with vertical side neck flasks, thermometers or bleed tubes can be angled into the bottom of the flask. Supplied complete with a compression cap and Viton o-ring.
Catalog No. 50-153-0990 Supplier Chemglass Life Sciences Supplier No. CG104429
Adapter, Thermometer, Inlet, 10¿ Offset, 29/26 Inner Joint. Adapter has a #7 Chem-Thread at the top for vacuum tight seal of plain stem thermometers or any other tube having an O.D. between 4 and 7mm. When used with vertical side neck flasks, thermometers or bleed tubes can be angled into the bottom of the flask. Supplied complete with a compression cap and Viton o-ring.