Learn More
Chemglass Life Sciences Adapter, Thermometer, 10° Offset, 24/40 Inner Joint
SDP
Catalog No. CG105401
Encompass_Preferred
Adapter, Thermometer, 10¿ Offset, 24/40 Inner Joint. Adapter has a 10/30 outer joint at the top for use with CG-3506 thermometers. When used with vertical side neck flasks, thermometers can be angled into the bottom of the flask. FOR THERMOMETERS SEE CG-3506-N.
Catalog No. CG105401 Supplier Chemglass Life Sciences Supplier No. CG105401
Adapter, Thermometer, 10¿ Offset, 24/40 Inner Joint. Adapter has a 10/30 outer joint at the top for use with CG-3506 thermometers. When used with vertical side neck flasks, thermometers can be angled into the bottom of the flask. FOR THERMOMETERS SEE CG-3506-N.