Learn More
Perkin Elmer US LLC Ball Joint Twist Type Torch Injector Support Adapter for ELAN DRCs
Catalog No. 509046871
Encompass_Preferred
This Injector Support Adapter is compatible with the 0.85 mm I.D. Quartz Ball Joint Injector (WE027030), 1.5 mm I.D. Quartz Ball Joint Injector (WE027005), and the 2.0 mm I.D. Quartz Ball Joint Injector (WE023948).
Catalog No. 50-904-6871 Supplier Perkin Elmer US LLC Supplier No. WE023951
This Injector Support Adapter is compatible with the 0.85 mm I.D. Quartz Ball Joint Injector (WE027030), 1.5 mm I.D. Quartz Ball Joint Injector (WE027005), and the 2.0 mm I.D. Quartz Ball Joint Injector (WE023948).