Learn More
Chemglass Life Sciences Adapter, Trap Connection, Airfree, Schlenk
SDP
Catalog No. 501530949
Encompass_Preferred
Adapter, Trap Connection, Airfree, Schlenk. Used to connect the AF-0300-13 manifold to the AF-0300-04 trap assembly. Adapter has a 0-20mm Chem-Cap valve with two #20 o-ring joints. Supplied complete with two #214 Viton o-rings.
Catalog No. 50-153-0949 Supplier Chemglass Life Sciences Supplier No. AF030014
Adapter, Trap Connection, Airfree, Schlenk. Used to connect the AF-0300-13 manifold to the AF-0300-04 trap assembly. Adapter has a 0-20mm Chem-Cap valve with two #20 o-ring joints. Supplied complete with two #214 Viton o-rings.