Learn More
Beckman Coulter 50 mL Conical Tube Adapter Not Compatible with BioSafe Lid; 7 Tubes per Adapter
Catalog No. NC2591184
Encompass_Preferred
Adapters for tubes in Beckman Coulter Life Sciences Allegra V-15R Benchtop VS 4.750 Swinging-Bucket Rotor
Catalog No. NC2591184 Supplier Beckman Coulter Supplier No. C63168
Adapters for tubes in Beckman Coulter Life Sciences Allegra V-15R Benchtop VS 4.750 Swinging-Bucket Rotor