Learn More
Chemglass Life Sciences Adapter, Type J (Black) Male Standard to Female Mini, STYLE B -ROUND PRONGS
SDP
Supplier:  Chemglass Life Sciences CG3499A11
Encompass_Preferred
Plain Hastelloy C-276 Thermocouple, 1/8" Diameter x 4" Long, Type "T", Standard Male Plug (OST). Thermocouples for use with Chemglass process reactors. The 4 long Hastelloy C-276 style is for use with the CG-1971 baffles and adapters. Detachable type thermocouples have a standard male plug (OST) and require a coiled extension cord for connection to the CG-3498 temperature monitor. The 4 long Hastelloy C-276 thermocouples are supplied with a 4ft FEP insulated cord having a standard male plug (OST).
Catalog No. 50-153-1628