Learn More
Chemglass Life Sciences Adapter, Type K (Yellow) Male Standard to Female Mini, STYLE B - ROUND PRONGS
SDP
Supplier:  Chemglass Life Sciences CG3499A04
Encompass_Preferred
Style B Thermocouple Transition Adapter, Type "K" (Yellow) Mini Male to Female Standard. Adapters provide quick and easy connections between standard (OST) and miniature (SMP) connector systems.
Catalog No. CG3499A04