Learn More
Chemglass Life Sciences Adapter, Universal, Inlet, 24/40 Inner Joint, #9 Chem-Thread, Hose Connection, Accepts O.D./7-9mm, O-Ring/110
SDP
Catalog No. CG104504
Encompass_Preferred
Adapter, Universal, Inlet, 24/40 Inner Joint, #9 Chem-Thread, Hose Connection, Accepts O.D./7-9mm, O-Ring/110. Adapter has the listed size of Chem-Thread at the top for a vacuum tight seal of a variety of tubing diameters. Will accept glass, metal or heavy wall plastic tubes having diameters between 7 to 18mm O.D. such as thermometer wells, gas dispersion tubes, plain stem funnels, etc. Serrated hose connection has an O.D. of 10mm at the largest serration. Adapters are supplied complete with a Celcon compression cap and Viton o-ring.
Catalog No. CG104504 Supplier Chemglass Life Sciences Supplier No. CG104504
Adapter, Universal, Inlet, 24/40 Inner Joint, #9 Chem-Thread, Hose Connection, Accepts O.D./7-9mm, O-Ring/110. Adapter has the listed size of Chem-Thread at the top for a vacuum tight seal of a variety of tubing diameters. Will accept glass, metal or heavy wall plastic tubes having diameters between 7 to 18mm O.D. such as thermometer wells, gas dispersion tubes, plain stem funnels, etc. Serrated hose connection has an O.D. of 10mm at the largest serration. Adapters are supplied complete with a Celcon compression cap and Viton o-ring.