Learn More
Cole-Parmer Cole-Parmer Adapter, U.S. standard meter to BNC electrode
Catalog No. 50633588 Shop All Cole-Parmer Products
Encompass_Preferred
BNC is the most common electrode connector, but some pH meters and electrodes come with different types of connectors.Order an adapter to ensure your meter and electrode are compatible.
Catalog No. 50-633-588 Supplier Cole-Parmer Supplier No. 0598325
BNC is the most common electrode connector, but some pH meters and electrodes come with different types of connectors.Order an adapter to ensure your meter and electrode are compatible.