Learn More
Chemglass Life Sciences Adapter, Vacuum Filtration, #1 Top Pluro Stopper, 14/20 Lower Inner, Fits Funnel Capacity 2 to 30mL
SDP
Catalog No. CG105214
Encompass_Preferred
Adapter, Vacuum Filtration, #1 Top Pluro Stopper, 14/20 Lower Inner, Fits Funnel Capacity 2 to 30mL. Adapters are used for reduced pressure filtration with plain stem Buchner funnels. Top is tooled to accept the listed size of pluro-stopper. Serrated hose connection has an O.D. of 10mm at the largest serration. FOR PLURO STOPPERS SEE CG-1401.
Catalog No. CG105214 Supplier Chemglass Life Sciences Supplier No. CG105214
Adapter, Vacuum Filtration, #1 Top Pluro Stopper, 14/20 Lower Inner, Fits Funnel Capacity 2 to 30mL. Adapters are used for reduced pressure filtration with plain stem Buchner funnels. Top is tooled to accept the listed size of pluro-stopper. Serrated hose connection has an O.D. of 10mm at the largest serration. FOR PLURO STOPPERS SEE CG-1401.