Learn More
Chemglass Life Sciences Adapter, Vacuum Take-Off, Long Stem, 24/40 Joint Size
SDP
Catalog No. CG1049E01
Encompass_Preferred
Adapter, Female Luer Lock, Vacuum, 1/4"-28 Thread, 14/20 Inner. Adapter has a 1/4” -28 thread that accepts a female luer lock adapter which provides a convenient way to connect solid phase extraction (SPE) cartridges, tubes, or syringes with a male luer lock fitting to a standard male taper inner joint. The adapter provides for easier sample concentration and clean-up while reducing the chances of cross-contamination. Hose connection is 10mm OD at the largest serration. Supplied complete with 1 each CG-1055-20; 1/4”-28 Female to Female luer lock adapter.
Catalog No. CG1049E01 Supplier Chemglass Life Sciences Supplier No. CG1049E01
Adapter, Female Luer Lock, Vacuum, 1/4"-28 Thread, 14/20 Inner. Adapter has a 1/4” -28 thread that accepts a female luer lock adapter which provides a convenient way to connect solid phase extraction (SPE) cartridges, tubes, or syringes with a male luer lock fitting to a standard male taper inner joint. The adapter provides for easier sample concentration and clean-up while reducing the chances of cross-contamination. Hose connection is 10mm OD at the largest serration. Supplied complete with 1 each CG-1055-20; 1/4”-28 Female to Female luer lock adapter.