Learn More
Chemglass Life Sciences Adapter, Vacuum Take-Off, Long Stem, 29/42 Joint Size
SDP
Catalog No. CG1049E02
Encompass_Preferred
Adapter, Vacuum Take-Off, Long Stem, 29/42 Joint Size. With standard taper top outer and lower inner joints with sealed in lower tube extending below inner joint. Tubing extends 100mm below the joint on the 14/20 and 19/22 sizes and 200mm on the 24/40, 29/26 and 29/42 sizes. Serrated hose connection having an O.D. of 10mm at the largest serration for connection to vacuum source.
Catalog No. CG1049E02 Supplier Chemglass Life Sciences Supplier No. CG1049E02
Adapter, Vacuum Take-Off, Long Stem, 29/42 Joint Size. With standard taper top outer and lower inner joints with sealed in lower tube extending below inner joint. Tubing extends 100mm below the joint on the 14/20 and 19/22 sizes and 200mm on the 24/40, 29/26 and 29/42 sizes. Serrated hose connection having an O.D. of 10mm at the largest serration for connection to vacuum source.