Learn More
Chemglass Life Sciences Adapter, Vacuum Take-Off, Style "A", 105°, 19/22 Joint Size
SDP
Supplier:  Chemglass Life Sciences CG105019
Encompass_Preferred
Adapter, Vacuum Take-Off, Style "A", 105¿, 19/22 Joint Size. With standard taper top outer and lower inner joints with sealed in drip tip. Adapter is bent at a 105¿ angle for use in vacuum distillation. Serrated hose connection has an O.D. of 10mm at the largest serration for connection to vacuum source.
Catalog No. CG105019