Learn More
Chemglass Life Sciences Adapter, Vacuum, 24/25 Outer to 24/25 Inner
SDP
Catalog No. 501539686
Encompass_Preferred
Adapter has a ground glass standard taper outer joint on top and bottom that provides a convenient leak free way to connect to the OP-6602-J 2L disposable long stem filter funnels and a standard taper flask. The unique design of the long stem filter funnel and vacuum adapter makes it unnecessary to clean after each use. Hose connection is 10mm OD at the largest serration.
Catalog No. 50-153-9686 Supplier Chemglass Life Sciences Supplier No. CG1049M24
Adapter has a ground glass standard taper outer joint on top and bottom that provides a convenient leak free way to connect to the OP-6602-J 2L disposable long stem filter funnels and a standard taper flask. The unique design of the long stem filter funnel and vacuum adapter makes it unnecessary to clean after each use. Hose connection is 10mm OD at the largest serration.