Learn More
Chemglass Life Sciences Adapter, Vacuum, 24/25 Outer to 45/50 Inner
SDP
Catalog No. 501539687
Encompass_Preferred
Adapter has a ground glass standard taper outer joint on top and bottom that provides a convenient leak free way to connect to the OP-6602-J 2L disposable long stem filter funnels and a standard taper flask. The unique design of the long stem filter funnel and vacuum adapter makes it unnecessary to clean after each use. Hose connection is 10mm OD at the largest serration.
Catalog No. 50-153-9687 Supplier Chemglass Life Sciences Supplier No. CG1049M45
Adapter has a ground glass standard taper outer joint on top and bottom that provides a convenient leak free way to connect to the OP-6602-J 2L disposable long stem filter funnels and a standard taper flask. The unique design of the long stem filter funnel and vacuum adapter makes it unnecessary to clean after each use. Hose connection is 10mm OD at the largest serration.