Learn More
Chemglass Life Sciences Adapter, Vacuum Distillation, 105°, Minum-Ware
SDP
Supplier:  Chemglass Life Sciences MW3601
Encompass_Preferred
Adapter, Vacuum Distillation, 105¿, Minum-Ware. Used in microscale vacuum distillations. Adapter has a top threaded outer joint and a lower inner joint with sealed in drip tip, all are 14/10. Side hose connection has an O.D. of 9mm at the middle serration. Supplied complete with open-top screw cap (MW-102-33) and Viton O-ring (MW-100-08).
Catalog No. 50-872-927