Learn More
Chemglass Life Sciences Adapter, Vacuum Filtration, 29/42, #3 Pluro Stopper, PTFE Stopcock, Accepts Buchner Funnel up to 150mL
SDP
Catalog No. 50872890
Encompass_Preferred
Adapter, Vacuum Filtration, 29/42, #3 Pluro Stopper, PTFE Stopcock, Accepts Buchner Funnel up to 150mL. Adapter used for reduced pressure filtrations using plain stem Buchner funnels. Stopcock permits control of vacuum source. Top of adapter is tooled to accept the listed size of pluro-stopper.NOTE: ADAPTER IS NOT SUPPLIED WITH RUBBER PLURO-STOPPER, SEE CG-1401
Catalog No. 50-872-890 Supplier Chemglass Life Sciences Supplier No. CG1053A03
Adapter, Vacuum Filtration, 29/42, #3 Pluro Stopper, PTFE Stopcock, Accepts Buchner Funnel up to 150mL. Adapter used for reduced pressure filtrations using plain stem Buchner funnels. Stopcock permits control of vacuum source. Top of adapter is tooled to accept the listed size of pluro-stopper.NOTE: ADAPTER IS NOT SUPPLIED WITH RUBBER PLURO-STOPPER, SEE CG-1401