Learn More
Chemglass Life Sciences Adapter, Vacuum Filtration, 14/20, #2 Pluro Stopper, PTFE Stopcock, Accepts Buchner Funnel up to 30mL
SDP
Catalog No. CG1053A01
Encompass_Preferred
Adapter, Vacuum Filtration, 14/20, #2 Pluro Stopper, PTFE Stopcock, Accepts Buchner Funnel up to 30mL. Adapter used for reduced pressure filtrations using plain stem Buchner funnels. Stopcock permits control of vacuum source. Top of adapter is tooled to accept the listed size of pluro-stopper.NOTE: ADAPTER IS NOT SUPPLIED WITH RUBBER PLURO-STOPPER, SEE CG-1401
Catalog No. CG1053A01 Supplier Chemglass Life Sciences Supplier No. CG1053A01
Adapter, Vacuum Filtration, 14/20, #2 Pluro Stopper, PTFE Stopcock, Accepts Buchner Funnel up to 30mL. Adapter used for reduced pressure filtrations using plain stem Buchner funnels. Stopcock permits control of vacuum source. Top of adapter is tooled to accept the listed size of pluro-stopper.NOTE: ADAPTER IS NOT SUPPLIED WITH RUBBER PLURO-STOPPER, SEE CG-1401