Learn More
Chemglass Life Sciences Adapter, Vacuum Take-Off, Style ''B'', 105, 19/22 Joint Size
SDP
Catalog No. 50872884
Encompass_Preferred
Adapter, Vacuum Take-Off, Style 'B', 105, 19/22 Joint Size. With standard taper top outer and lower inner joints with sealed in drip tip. Adapter is bent at a 105 angle for use in vacuum distillation. Serrated hose connection has an O.D. of 10mm at the largest serration for connection to vacuum source.
Catalog No. 50-872-884 Supplier Chemglass Life Sciences Supplier No. CG105071
Adapter, Vacuum Take-Off, Style 'B', 105, 19/22 Joint Size. With standard taper top outer and lower inner joints with sealed in drip tip. Adapter is bent at a 105 angle for use in vacuum distillation. Serrated hose connection has an O.D. of 10mm at the largest serration for connection to vacuum source.