Learn More
Chemglass Life Sciences Adapter, Vacuum Take-Off, Style "B", 105°, 14/20 Joint Size
SDP
Catalog No. 50872883
Encompass_Preferred
Adapter, Vacuum Take-Off, Style "B", 105¿, 14/20 Joint Size. With standard taper top outer and lower inner joints with sealed in drip tip. Adapter is bent at a 105¿ angle for use in vacuum distillation. Serrated hose connection has an O.D. of 10mm at the largest serration for connection to vacuum source.
Catalog No. 50-872-883 Supplier Chemglass Life Sciences Supplier No. CG105070
Adapter, Vacuum Take-Off, Style "B", 105¿, 14/20 Joint Size. With standard taper top outer and lower inner joints with sealed in drip tip. Adapter is bent at a 105¿ angle for use in vacuum distillation. Serrated hose connection has an O.D. of 10mm at the largest serration for connection to vacuum source.