Learn More
Chemglass Life Sciences Adapter, Vacuum Take-Off, Style "B", 105°, 24/40 Joint Size
SDP
Catalog No. 50872885
Encompass_Preferred
Adapter, Vacuum Take-Off, Style "B", 105¿, 24/40 Joint Size. With standard taper top outer and lower inner joints with sealed in drip tip. Adapter is bent at a 105¿ angle for use in vacuum distillation. Serrated hose connection has an O.D. of 10mm at the largest serration for connection to vacuum source.
Catalog No. 50-872-885 Supplier Chemglass Life Sciences Supplier No. CG105072
Adapter, Vacuum Take-Off, Style "B", 105¿, 24/40 Joint Size. With standard taper top outer and lower inner joints with sealed in drip tip. Adapter is bent at a 105¿ angle for use in vacuum distillation. Serrated hose connection has an O.D. of 10mm at the largest serration for connection to vacuum source.