Learn More
Chemglass Life Sciences Adapter, Vacuum Take-Off, Vertical, 29/42 Joint Size
SDP
Catalog No. 50872874
Encompass_Preferred
Adapter, Vacuum Take-Off, Vertical, 29/42 Joint Size. With standard taper top outer and lower inner joints with sealed in drip tip. Serrated hose connection has an O.D. of 10mm at the largest serration for connection to vacuum source.
Catalog No. 50-872-874 Supplier Chemglass Life Sciences Supplier No. CG104902
Adapter, Vacuum Take-Off, Vertical, 29/42 Joint Size. With standard taper top outer and lower inner joints with sealed in drip tip. Serrated hose connection has an O.D. of 10mm at the largest serration for connection to vacuum source.