Learn More
IDEX Health & Science Upchurch ADAPTER X-LONG 1/4-28 TO 10-32
Catalog No. NC1919467
Encompass_Preferred
Idex Threaded Port Adapters, Diameter Inner: 0.03 in., PEEK, Thread Size: 10-32 to 1/4-28, Swept Volume: 6.7 uL, Pressure: 1000 psi
Catalog No. NC1919467 Supplier IDEX Health & Science Upchurch Supplier No. P654
Idex Threaded Port Adapters, Diameter Inner: 0.03 in., PEEK, Thread Size: 10-32 to 1/4-28, Swept Volume: 6.7 uL, Pressure: 1000 psi