Learn More
Chemglass Life Sciences Adapter, Inlet, 29/42 Outer Joint, 100mm Height
SDP
Catalog No. CG101302
Encompass_Preferred
Adapter, Inlet, 29/42 Outer Joint, 100mm Height. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration.
Catalog No. CG101302 Supplier Chemglass Life Sciences Supplier No. CG101302
Adapter, Inlet, 29/42 Outer Joint, 100mm Height. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection has an O.D. of 10mm at the largest serration.