Learn More
Thomas Scientific Adapters for 0.2mL tubes
SDP
Catalog No. 501458807
Encompass
Unlike traditional mini centrifuges, the MyFuge 12 eliminates the need to change rotors when switching between microtubes and PCR strips. The included, unique COMBI-Rotor™ is all that is required for running 12 microtubes and 4 PCR strips simultaneously. With a fixed speed that produces 2,000 xg, this centrifuge is perfect for quick spin downs. Simply close the lid and the unit quickly ramps up to 5500 rpm. Open the lid, and the unit quickly decelerates for removal of samples. At just 5.5 inches wide, the MyFuge 12 truly is a personal centrifuge with unmatched capacity and flexibility. COMBI-Rotor™, for tubes and strips Twice the capacity of traditional mini centrifuges Near silent operation Starts & stops with opening/closing of the lid Conserves valuable bench space
Catalog No. 50-145-8807 Supplier Thomas Scientific Supplier No. 1185Z36
Unlike traditional mini centrifuges, the MyFuge 12 eliminates the need to change rotors when switching between microtubes and PCR strips. The included, unique COMBI-Rotor™ is all that is required for running 12 microtubes and 4 PCR strips simultaneously. With a fixed speed that produces 2,000 xg, this centrifuge is perfect for quick spin downs. Simply close the lid and the unit quickly ramps up to 5500 rpm. Open the lid, and the unit quickly decelerates for removal of samples. At just 5.5 inches wide, the MyFuge 12 truly is a personal centrifuge with unmatched capacity and flexibility. COMBI-Rotor™, for tubes and strips Twice the capacity of traditional mini centrifuges Near silent operation Starts & stops with opening/closing of the lid Conserves valuable bench space