Learn More
Thermo Scientific™ Adapters for Single Blood Bag Bucket with Filter Pack for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
GSA_VA
Catalog No. 75003842
Change view
Click to view available options
Type:
Blood Bag Adapter
Filter Holder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
75-003-842 Blood Bag Adapter
75-003-859 Filter Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75-003-842 Supplier Thermo Scientific™ Supplier No. 75003842
Only null left
Add to Cart
Add to Cart
Accommodate 450–550mL Triple, Quad or Quint blood bags with additive solution in single blood bag buckets with filter pack for Thermo Scientific™ HAEMAFlex™6/8 rotors.
Thermo Scientific™ blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.
Select an adapter for 450–550mL Triple, Quad or Quint blood bag with additive solution.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Blood Bag Adapter
Capacity 6/8 x 550 mL
For Use With (Equipment) Single blood bag bucket with filter pack
Description 450-550mL Triple, Quad or Quint Blood Bag with Additive Solution Adapter (set of 2)