Learn More
Chemglass Life Sciences Adapter, Stainless Steel, 30mm Tool-Free O-ring Flange to M16 Male Thread, 90°
SDP
Catalog No. 50143630
Encompass_Preferred
Adapter, Stainless Steel, 30mm Tool-Free O-ring Flange to M16 Male Thread, 90¿. Stainless steel transition adapter from 30mm Tool-Free Glass Flange on the inlet/outlet of the reactor to M16. Tool-Free 30mm O-ring Flange is used to connect fluid transfer hose to the vessel. Adapters are supplied with PTFE Split Ring, aluminum lock collar and a fluorosilicate o-ring.
Catalog No. 50-143-630 Supplier Chemglass Life Sciences Supplier No. CG1969V10
Adapter, Stainless Steel, 30mm Tool-Free O-ring Flange to M16 Male Thread, 90¿. Stainless steel transition adapter from 30mm Tool-Free Glass Flange on the inlet/outlet of the reactor to M16. Tool-Free 30mm O-ring Flange is used to connect fluid transfer hose to the vessel. Adapters are supplied with PTFE Split Ring, aluminum lock collar and a fluorosilicate o-ring.