Learn More
Chemglass Life Sciences Adapter, Connecting, 14/20 Top Outer, 10/30 Lower Inner, Enlarging
SDP
Catalog No. CG100228
Encompass_Preferred
Adapter, Connecting, 14/20 Top Outer, 10/30 Lower Inner, Enlarging. Used to connect dissimilar standard taper joint sizes. Lower inner joint is smaller in size than the top outer joint.
Catalog No. CG100228 Supplier Chemglass Life Sciences Supplier No. CG100228
Adapter, Connecting, 14/20 Top Outer, 10/30 Lower Inner, Enlarging. Used to connect dissimilar standard taper joint sizes. Lower inner joint is smaller in size than the top outer joint.