Learn More
Chemglass Life Sciences Adapter, PTFE, 1/4in Thermocouple, 24/40 Inner Joint
SDP
Catalog No. CG1048A05
Encompass_Preferred
Adapter, PTFE, 1/4in Thermocouple, 24/40 Inner Joint. PTFE adapter for use with either 1/8 or 1/4 thermocouples. Adapter has an outer Viton o-ring to form a grease-free, airtight seal between the adapter and the listed size of outer joint. The compression nut works in conjunction with the upper Viton to provide a airtight seal between the adapter and thermocouple. Adapter design permits use with CG-145 Keck clips.
Catalog No. CG1048A05 Supplier Chemglass Life Sciences Supplier No. CG1048A05
Adapter, PTFE, 1/4in Thermocouple, 24/40 Inner Joint. PTFE adapter for use with either 1/8 or 1/4 thermocouples. Adapter has an outer Viton o-ring to form a grease-free, airtight seal between the adapter and the listed size of outer joint. The compression nut works in conjunction with the upper Viton to provide a airtight seal between the adapter and thermocouple. Adapter design permits use with CG-145 Keck clips.