Learn More
V&P Scientific Inc TUBE INSERT, Clear Acrylic Cylinder with an Offset Hole
Catalog No. NC0547715
Encompass
TUBE INSERT, Clear Acrylic Cylinder with an Offset Hole, Holds 1 x 15mL Conical Centrifuge Tube, used with VP 772FB-1, can also hold 13mL Standard Plastic and 10mL Glass Tubes
Catalog No. NC0547715 Supplier V&P Scientific Inc Supplier No. VP 772FB1A
TUBE INSERT, Clear Acrylic Cylinder with an Offset Hole, Holds 1 x 15mL Conical Centrifuge Tube, used with VP 772FB-1, can also hold 13mL Standard Plastic and 10mL Glass Tubes