Learn More
Glass Expansion Adaptor for D-Torch B13
SDP
Catalog No. 502703120
Encompass_Preferred
Adaptor for D-Torch B13. Compatible with Glass Expansion D-Torch (P/N 501907035, 30-808-3614) for Spectro (Ametek) Arcos II SOP, Blue SOP & Green SOP/DSOI ICP-OES. Provides a 13mm male ball joint. Replacement O-Rings are P/Ns 70-V-011 and 70-VT-012. Use O-ring removal tool 50-100-1636.For D-Torch Adaptor with 19mm male ball joint P/N 31-808-3615.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3120 Supplier Glass Expansion Supplier No. 318083596
Adaptor for D-Torch B13. Compatible with Glass Expansion D-Torch (P/N 501907035, 30-808-3614) for Spectro (Ametek) Arcos II SOP, Blue SOP & Green SOP/DSOI ICP-OES. Provides a 13mm male ball joint. Replacement O-Rings are P/Ns 70-V-011 and 70-VT-012. Use O-ring removal tool 50-100-1636.For D-Torch Adaptor with 19mm male ball joint P/N 31-808-3615.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001