Learn More
Glass Expansion Adaptor for D-Torch B19
SDP
Catalog No. 502703119
Encompass_Preferred
Adaptor for D-Torch B19. Compatible with Glass Expansion D-Torch (P/N 501907035, 30-808-3614) for Spectro (Ametek) Arcos II SOP, Blue SOP & Green SOP/DSOI ICP-OES. Provides a 19mm male ball joint. Replacement O-Rings are P/Ns 70-V-011 and 70-VT-012. Use O-ring removal tool 50-100-1636.For D-Torch Adaptor with 13mm male ball joint P/N 31-808-3596.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3119 Supplier Glass Expansion Supplier No. 318083651
Adaptor for D-Torch B19. Compatible with Glass Expansion D-Torch (P/N 501907035, 30-808-3614) for Spectro (Ametek) Arcos II SOP, Blue SOP & Green SOP/DSOI ICP-OES. Provides a 19mm male ball joint. Replacement O-Rings are P/Ns 70-V-011 and 70-VT-012. Use O-ring removal tool 50-100-1636.For D-Torch Adaptor with 13mm male ball joint P/N 31-808-3596.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001